This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02834898_m1
See other MSS51 GE Assays ›
Gene Symbol:
MSS51
Gene Name:
MSS51 mitochondrial translational activator
Gene Aliases:
ZMYND17
Chromosome Location:
Chr.9: 64281761 - 64293825 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02834898_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSS51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015147379.2 XP_015002865.2 2715 93
XM_015147380.2 XP_015002866.2 2871 93
XM_015147381.2 XP_015002867.2 2791 93
GenBank mRNA JU479921.1 - 549 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0