This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02835379_m1
See other ATF3 GE Assays ›
Gene Symbol:
ATF3
Gene Name:
activating transcription factor 3
Gene Aliases:
EGK_01674
Chromosome Location:
Chr.1: 70055871 - 70109651 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02835379_m1
Size
Availability Inventoried
Catalog # 4331182
Price 272.00
Your Price
Online offer: 256.65
256.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266010.1 NP_001252939.1 425 144
XM_015116707.2 XP_014972193.1 504 144
XM_015116716.2 XP_014972202.2 1188 144
XM_028844193.1 XP_028700026.1 4677 144
GenBank mRNA JU322209.1 - 425 144

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-m9fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline