This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02835659_m1
See other MMP28 GE Assays ›
Gene Symbol:
MMP28
Gene Name:
matrix metallopeptidase 28
Gene Aliases:
-
Chromosome Location:
Chr.16: 32112299 - 32154957 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02835659_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMP28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278395.2 NP_001265324.1 326 63
NM_001278396.2 NP_001265325.1 326 63
XM_015119023.2 XP_014974509.1 519 63
XM_015119024.2 XP_014974510.1 913 63
XM_028835451.1 XP_028691284.1 261 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028835451.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

metalloprotease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0