This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02836366_m1
See other TTPAL GE Assays ›
Gene Symbol:
TTPAL
Gene Name:
alpha tocopherol transfer protein like
Gene Aliases:
EGK_02385
Chromosome Location:
Chr.10: 79701477 - 79716517 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02836366_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTPAL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001265955.1 NP_001252884.1 548 147
XM_015148844.2 XP_015004330.1 1776 147
XM_015148845.2 XP_015004331.1 1030 147
XM_015148848.2 XP_015004334.1 1030 147
XM_028827349.1 XP_028683182.1 1779 147
GenBank mRNA JU336364.1 - 548 147
JU472731.1 - 547 147
JV723251.1 - 549 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transfer/carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0