This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02837159_m1
See other PTPN20 GE Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase non-receptor type 20
Gene Aliases:
EGK_19635, PTPN20A
Chromosome Location:
Chr.9: 49239796 - 49340960 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02837159_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001109336.4 XP_001109336.1 644 111
XM_015146962.2 XP_015002448.1 1132 111
XM_015146965.2 XP_015002451.1 2390 111
XM_015146968.2 XP_015002454.1 554 111
XM_015146970.2 XP_015002456.1 465 111
XM_015146971.2 XP_015002457.1 376 111
XM_028826354.1 XP_028682187.1 1094 111
XM_028826355.1 XP_028682188.1 1195 111
XM_028826356.1 XP_028682189.1 697 111
XM_028826357.1 XP_028682190.1 1002 111
XM_028826358.1 XP_028682191.1 1082 111
XM_028826359.1 XP_028682192.1 554 111

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015146970.2, XM_015146971.2, XM_028826359.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0