This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02847539_m1
See other FCGRT GE Assays ›
Gene Symbol:
FCGRT LOC718859
Gene Name:
Fc fragment of IgG receptor and transporter
Fc fragment of IgG receptor and transporter
Gene Aliases:
-
EGK_10892, FCGRT
Chromosome Location:
Chr.19: 49475672 - 49492034 on Build Mmul_10
Chromosome Location:
Chr.19: 22694 - 38307 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh02847539_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FCGRT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028839697.1 XP_028695530.1 1435 174
XM_028839698.1 XP_028695531.1 1481 174
Gene Symbol: LOC718859
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257520.1 NP_001244449.1 175
GenBank mRNA JU322969.1 - 175
JU474694.1 - 175

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline