This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02848597_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 59867378 - 60026998 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02848597_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001261000.1
NP_001247929.1
569
78
XM_015117598.2
XP_014973084.1
973
78
XM_015117599.2
XP_014973085.1
454
78
XM_015117600.2
XP_014973086.1
973
78
XM_015117601.2
XP_014973087.1
970
78
XM_015117602.2
XP_014973088.1
973
78
XM_015117603.2
XP_014973089.1
973
78
XM_015117604.2
XP_014973090.1
769
78
XM_015117605.2
XP_014973091.1
589
78
XM_015117606.2
XP_014973092.1
882
78
XM_015117607.2
XP_014973093.1
882
78
XM_028834315.1
XP_028690148.1
630
78
XM_028834316.1
XP_028690149.1
680
78
XM_028834317.1
XP_028690150.1
970
78
XM_028834318.1
XP_028690151.1
680
78
XM_028834319.1
XP_028690152.1
561
78
XM_028834320.1
XP_028690153.1
563
78
XM_028834321.1
XP_028690154.1
473
78
XM_028834322.1
XP_028690155.1
360
78
XM_028834324.1
XP_028690157.1
536
78
XM_028834325.1
XP_028690158.1
589
78
XM_028834326.1
XP_028690159.1
882
78
XM_028834327.1
XP_028690160.1
564
78
GenBank mRNA
JU320297.1
-
560
78
JV046385.1
-
555
78
JV046386.1
-
569
78
JV497440.1
-
606
78
JV636009.1
-
510
78
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
translation initiation factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0