Assay Details
| Gene Symbol: MAP3K21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265676.1 | NP_001252605.1 | 1549 | 124 | ||
| XM_028844719.1 | XP_028700552.1 | 1620 | 124 | |||
| GenBank mRNA | JV048457.1 | - | 1429 | 124 | ||
| JV636412.1 | - | 1549 | 124 | |||
More Information
Set Membership: |
|

MAP3K21
Genomic Map