This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02853590_m1
See other SDSL GE Assays ›
Gene Symbol:
SDSL
Gene Name:
serine dehydratase like
Gene Aliases:
-
Chromosome Location:
Chr.11: 113133379 - 113155024 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02853590_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDSL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015152888.2 XP_015008374.1 618 71
XM_028829941.1 XP_028685774.1 435 71
XM_028829942.1 XP_028685775.1 894 71
XM_028829943.1 XP_028685776.1 435 71
XM_028829944.1 XP_028685777.1 903 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw9fd:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0