This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02862901_m1
See other PEMT GE Assays ›
Gene Symbol:
PEMT
Gene Name:
phosphatidylethanolamine N-methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.16: 17330577 - 17416283 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02862901_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PEMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001090659.4 XP_001090659.1 396 113
XM_001090779.4 XP_001090779.1 426 113
XM_015118738.2 XP_014974224.1 349 113
XM_015118739.2 XP_014974225.1 437 113
GenBank mRNA JU332497.1 - 385 113
JV045673.1 - 377 113
JV568869.1 - 383 113
JV637736.1 - 377 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0