This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02864750_m1
See other SLCO3A1 GE Assays ›
Gene Symbol:
SLCO3A1
Gene Name:
solute carrier organic anion transporter family member 3A1
Gene Aliases:
-
Chromosome Location:
Chr.7: 70914495 - 71233198 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02864750_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO3A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261477.1 NP_001248406.1 1518 64
XM_015143057.2 XP_014998543.2 1904 64
XM_015143059.2 XP_014998545.2 1904 64
XM_015143060.2 XP_014998546.2 1904 64
XM_028850432.1 XP_028706265.1 1904 64
XM_028850433.1 XP_028706266.1 1904 64
GenBank mRNA JU322154.1 - 1518 64
JU334074.1 - 1518 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0