This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02864759_m1
See other RACGAP1 GE Assays ›
Gene Symbol:
RACGAP1
Gene Name:
Rac GTPase activating protein 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 49596533 - 49632504 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02864759_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RACGAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266442.1 NP_001253371.1 1459 94
XM_015151631.2 XP_015007117.1 1586 94
XM_015151632.2 XP_015007118.1 1463 94
XM_015151633.2 XP_015007119.1 1432 94
XM_015151634.2 XP_015007120.1 1361 94
XM_028828953.1 XP_028684786.1 1457 94
XM_028828954.1 XP_028684787.1 1405 94
GenBank mRNA JU334170.1 - 1350 94
JU473254.1 - 1463 94
JU473255.1 - 1459 94
JV703539.1 - 1471 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0