This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02869564_m1
See other PPWD1 GE Assays ›
Gene Symbol:
PPWD1
Gene Name:
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases:
EGK_16537
Chromosome Location:
Chr.6: 64130310 - 64156072 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02869564_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPWD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266376.1 NP_001253305.1 1541 74
XM_015140109.2 XP_014995595.1 1495 74
XM_015140110.2 XP_014995596.1 1501 74
XM_015140111.2 XP_014995597.1 1593 74
XM_015140112.2 XP_014995598.1 1112 74
XM_028849094.1 XP_028704927.1 1442 74
GenBank mRNA JU333675.1 - 1541 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0