This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02870715_m1
See other NUDT13 GE Assays ›
Gene Symbol:
NUDT13
Gene Name:
nudix hydrolase 13
Gene Aliases:
-
Chromosome Location:
Chr.9: 64586031 - 64626835 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02870715_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015147389.2 XP_015002875.2 418 99
XM_015147392.2 XP_015002878.2 247 99
XM_015147393.2 XP_015002879.2 247 99
XM_015147394.2 XP_015002880.2 368 99
XM_015147395.2 XP_015002881.1 418 99
XM_015147396.2 XP_015002882.2 247 99
XM_028826488.1 XP_028682321.1 486 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015147394.2, XM_015147395.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0