This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02871949_m1
See other JKAMP GE Assays ›
Gene Symbol:
JKAMP
Gene Name:
JNK1/MAPK8 associated membrane protein
Gene Aliases:
C7H14orf100
Chromosome Location:
Chr.7: 121583276 - 121605781 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02871949_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JKAMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260993.1 NP_001247922.1 315 88
XM_015143763.2 XP_014999249.1 373 88
XM_015143764.2 XP_014999250.1 423 88
GenBank mRNA JU328135.1 - 140 88
JU471962.1 - 315 88
JV636210.1 - 265 88

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0