This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02874617_m1
See other ARFGAP1 GE Assays ›
Gene Symbol:
ARFGAP1
Gene Name:
ADP ribosylation factor GTPase activating protein 1
Gene Aliases:
EGK_02194
Chromosome Location:
Chr.10: 98482999 - 98501020 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02874617_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARFGAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261226.1 NP_001248155.1 730 103
XM_015148572.2 XP_015004058.2 767 103
XM_015148574.2 XP_015004060.2 1481 103
XM_015148575.2 XP_015004061.2 767 103
XM_015148578.2 XP_015004064.1 767 103
XM_015148579.2 XP_015004065.1 767 103
XM_015148581.2 XP_015004067.1 289 103
XM_028827456.1 XP_028683289.1 1481 103
XM_028827457.1 XP_028683290.1 646 103
XM_028827458.1 XP_028683291.1 1474 103
XM_028827459.1 XP_028683292.1 1475 103
XM_028827460.1 XP_028683293.1 1474 103
XM_028827461.1 XP_028683294.1 1474 103
XM_028827462.1 XP_028683295.1 1474 103
XM_028827463.1 XP_028683296.1 646 103
XM_028827464.1 XP_028683297.1 646 103
GenBank mRNA JU323837.1 - 743 103
JU473403.1 - 663 103
JV047613.1 - 730 103
JV527809.1 - 657 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0