This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02876613_m1
See other GPR137 GE Assays ›
Gene Symbol:
GPR137
Gene Name:
G protein-coupled receptor 137
Gene Aliases:
-
Chromosome Location:
Chr.14: 10252793 - 10273617 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02876613_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR137
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261715.1 NP_001248644.1 753 84
XM_015113729.2 XP_014969215.1 1048 84
XM_015113730.2 XP_014969216.1 989 84
XM_028832814.1 XP_028688647.1 713 84
XM_028832815.1 XP_028688648.1 894 84
XM_028832816.1 XP_028688649.1 1323 84
XM_028832817.1 XP_028688650.1 989 84
XM_028832818.1 XP_028688651.1 1048 84
XM_028832819.1 XP_028688652.1 989 84
XM_028832820.1 XP_028688653.1 713 84
GenBank mRNA JU320461.1 - 771 84
JU476449.1 - 757 84
JV044851.1 - 953 84
JV044852.1 - 753 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0