This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02877052_m1
See other EIF2B3 GE Assays ›
Gene Symbol:
EIF2B3
Gene Name:
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases:
EGK_00691
Chromosome Location:
Chr.1: 179430370 - 179576343 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02877052_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF2B3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260654.1 NP_001247583.1 1276 79
XM_015138005.2 XP_014993491.1 1343 79
XM_028843036.1 XP_028698869.1 1175 79
GenBank mRNA JU321084.1 - 1306 79
JU477677.1 - 1276 79
JV048601.1 - 1276 79
JV664641.1 - 1077 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0