This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02877510_m1
See other TMIGD3 GE Assays ›
Gene Symbol:
TMIGD3
Gene Name:
transmembrane and immunoglobulin domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 112627498 - 112715269 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02877510_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMIGD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001302780.1 NP_001289709.1 1333 126
NM_001302781.1 NP_001289710.1 726 126

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0