This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02878577_m1
See other PPM1A GE Assays ›
Gene Symbol:
PPM1A
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent 1A
Gene Aliases:
-
Chromosome Location:
Chr.7: 122343294 - 122395709 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02878577_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPM1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001096358.4 XP_001096358.2 1360 154
XM_015143816.2 XP_014999302.1 1796 154
XM_015143817.2 XP_014999303.1 1888 154
XM_015143818.2 XP_014999304.1 1743 154
XM_015143819.2 XP_014999305.1 1173 154
XM_028851590.1 XP_028707423.1 1532 154
GenBank mRNA JV638234.1 - 1362 154

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0