This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02880417_m1
See other PCED1A GE Assays ›
Gene Symbol:
PCED1A
Gene Name:
PC-esterase domain containing 1A
Gene Aliases:
FAM113A
Chromosome Location:
Chr.10: 63663146 - 63669071 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02880417_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCED1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194663.3 NP_001181592.3 684 93
XM_015149355.2 XP_015004841.2 942 93
XM_015149356.2 XP_015004842.2 832 93
XM_015149357.2 XP_015004843.2 598 93
GenBank mRNA JU321750.1 - 908 93
JU473686.1 - 900 93
JV045252.1 - 791 93
JV658975.1 - 915 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline