This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02881129_m1
See other ZNF343 GE Assays ›
Gene Symbol:
ZNF343
Gene Name:
zinc finger protein 343
Gene Aliases:
EGK_02596
Chromosome Location:
Chr.10: 63998709 - 64020652 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02881129_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF343
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260644.1 NP_001247573.1 351 159
XM_015149350.2 XP_015004836.1 408 159
XM_028827396.1 XP_028683229.1 408 159
XM_028827397.1 XP_028683230.1 408 159
GenBank mRNA JU323246.1 - 351 159
JU473241.1 - 351 159
JV637962.1 - 358 159

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline