This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02881410_m1
See other RIC3 GE Assays ›
Gene Symbol:
RIC3
Gene Name:
RIC3 acetylcholine receptor chaperone
Gene Aliases:
-
Chromosome Location:
Chr.14: 57518717 - 57576484 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02881410_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261177.1 NP_001248106.1 456 131
XM_015114745.2 XP_014970231.1 545 131
XM_028832639.1 XP_028688472.1 545 131
XM_028832640.1 XP_028688473.1 594 131
XM_028832641.1 XP_028688474.1 507 131
XM_028832642.1 XP_028688475.1 594 131
XM_028832643.1 XP_028688476.1 288 131
XM_028832644.1 XP_028688477.1 257 131
XM_028832645.1 XP_028688478.1 545 131
GenBank mRNA JU336386.1 - 460 131
JV048253.1 - 456 131
JV723342.1 - 463 131

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0