This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02882070_m1
See other CBLL1 GE Assays ›
Gene Symbol:
CBLL1
Gene Name:
Cbl proto-oncogene like 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 134062759 - 134081389 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02882070_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CBLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260727.2 NP_001247656.1 482 137
XM_028844855.1 XP_028700688.1 334 137
XM_028844857.1 XP_028700690.1 191 137
GenBank mRNA JU320760.1 - 266 137
JU477254.1 - 487 137

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028844857.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0