This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02884807_m1
See other LRTOMT GE Assays ›
Gene Symbol:
LRTOMT
Gene Name:
leucine rich transmembrane and O-methyltransferase domain containing
Gene Aliases:
EGK_06641
Chromosome Location:
Chr.14: 65031402 - 65061492 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02884807_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRTOMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136102.1 NP_001129574.1 614 94
NM_001199354.1 NP_001186283.1 431 94
XM_015115045.2 XP_014970531.1 845 94
XM_015115046.2 XP_014970532.1 862 94
XM_015115047.2 XP_014970533.1 440 94
XM_015115048.2 XP_014970534.1 662 94
XM_015115049.2 XP_014970535.1 429 94
XM_015115050.2 XP_014970536.1 847 94
XM_028832845.1 XP_028688678.1 991 94
GenBank mRNA EU627076.1 - 431 94
EU627077.1 - 431 94
EU627078.1 - 614 94
EU627079.1 - 431 94
JV046448.1 - 340 94
JV635861.1 - 487 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001136102.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline