This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02885418_m1
See other METTL6 GE Assays ›
Gene Symbol:
METTL6
Gene Name:
methyltransferase like 6
Gene Aliases:
EGK_12118
Chromosome Location:
Chr.2: 32240017 - 32285898 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02885418_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266134.2 NP_001253063.1 414 79
XM_015132160.2 XP_014987646.1 480 79
XM_015132162.2 XP_014987648.1 468 79
XM_015132163.2 XP_014987649.2 481 79
XM_015132164.2 XP_014987650.1 481 79
XM_015132167.2 XP_014987653.1 481 79
XM_015132168.2 XP_014987654.1 481 79
XM_015132169.2 XP_014987655.1 483 79
XM_028843086.1 XP_028698919.1 481 79
GenBank mRNA JU323392.1 - 414 79
JU472818.1 - 454 79
JV044119.1 - 416 79
JV642439.1 - 469 79

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0