This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02888989_m1
See other DTX3 GE Assays ›
Gene Symbol:
DTX3
Gene Name:
deltex E3 ubiquitin ligase 3
Gene Aliases:
-
Chromosome Location:
Chr.11: 57181762 - 57187064 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02888989_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DTX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015152133.2 XP_015007619.1 1242 74
XM_015152135.2 XP_015007621.1 1256 74
XM_015152137.2 XP_015007623.1 1594 74
XM_028829674.1 XP_028685507.1 1001 74
XM_028829675.1 XP_028685508.1 1882 74
XM_028829676.1 XP_028685509.1 1020 74
XM_028829677.1 XP_028685510.1 1897 74
GenBank mRNA JU320050.1 - 891 74
JU474592.1 - 893 74
JV046374.1 - 1025 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0