This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02890369_m1
See other NHS GE Assays ›
Gene Symbol:
NHS
Gene Name:
NHS actin remodeling regulator
Gene Aliases:
-
Chromosome Location:
Chr.X: 17109052 - 17475237 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02890369_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NHS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001104229.4 XP_001104229.2 1655 75
XM_001104388.4 XP_001104388.1 1592 75
XM_015127014.2 XP_014982500.1 967 75
XM_015127015.2 XP_014982501.1 904 75
XM_015127017.2 XP_014982503.1 1064 75
XM_028841985.1 XP_028697818.1 1813 75
GenBank mRNA JV508031.1 - 75 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0