This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02890442_m1
See other HACD2 GE Assays ›
Gene Symbol:
HACD2
Gene Name:
3-hydroxyacyl-CoA dehydratase 2
Gene Aliases:
PTPLB
Chromosome Location:
Chr.2: 152727687 - 152826545 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02890442_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261129.1 NP_001248058.1 521 70
XM_028843389.1 XP_028699222.1 547 70
XM_028843390.1 XP_028699223.1 325 70
XM_028843391.1 XP_028699224.1 568 70
XM_028843392.1 XP_028699225.1 231 70
GenBank mRNA JU327265.1 - 496 70
JV044709.1 - 521 70
JV639352.1 - 517 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0