This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02890509_m1
See other PRSS55 GE Assays ›
Gene Symbol:
PRSS55
Gene Name:
serine protease 55
Gene Aliases:
EGK_18697
Chromosome Location:
Chr.8: 10023782 - 10124833 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02890509_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001089204.4 XP_001089204.1 796 98
XM_015144766.2 XP_015000252.1 888 98
XM_028852332.1 XP_028708165.1 463 98
XM_028852333.1 XP_028708166.1 889 98
XM_028852334.1 XP_028708167.1 573 98
XM_028852335.1 XP_028708168.1 461 98
XM_028852336.1 XP_028708169.1 885 98
XM_028852337.1 XP_028708170.1 446 98
XM_028852338.1 XP_028708171.1 1105 98
GenBank mRNA JU325212.1 - 197 98
JV459729.1 - 55 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline