Assay Details
| Gene Symbol: CYP1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266868.1 | NP_001253797.1 | 364 | 103 | ||
| GenBank mRNA | JU474459.1 | - | 364 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266868.1 transcripts.
Set Membership: |
|

CYP1B1
Genomic Map