This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02913947_s1
See other SIAH1 GE Assays ›
Gene Symbol:
SIAH1
Gene Name:
siah E3 ubiquitin protein ligase 1
Gene Aliases:
EGK_12755
Chromosome Location:
Chr.20: 35419607 - 35510756 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02913947_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIAH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260838.2 NP_001247767.1 1995 168
XM_015126059.2 XP_014981545.1 1916 168
XM_015126060.2 XP_014981546.1 1961 168
XM_015126061.2 XP_014981547.1 1991 168
XM_015126065.1 XP_014981551.1 1765 168
XM_028840589.1 XP_028696422.1 1815 168
XM_028840590.1 XP_028696423.1 2049 168
GenBank mRNA JU347249.1 - 1832 168
JV043508.1 - 1998 168

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5lprg:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0