This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02914763_g1
See other SOCS2 GE Assays ›
Gene Symbol:
SOCS2
Gene Name:
suppressor of cytokine signaling 2
Gene Aliases:
EGK_04058
Chromosome Location:
Chr.11: 93367615 - 93374511 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02914763_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOCS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194762.2 NP_001181691.1 622 96
XM_001106151.4 XP_001106151.2 662 96
XM_002798726.3 XP_002798772.1 938 96
XM_002798729.3 XP_002798775.1 744 96
XM_015152399.2 XP_015007885.1 607 96
XM_015152401.2 XP_015007887.1 444 96
GenBank mRNA JU472267.1 - 359 96
JV045775.1 - 295 96

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

kinase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline