This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02982736_g1
See other LOC701715 GE Assays ›
Gene Symbol:
LOC701715
Gene Name:
pulmonary surfactant-associated protein A
Gene Aliases:
SFTPA2
Chromosome Location:
Chr.9: 58275870 - 58285786 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02982736_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC701715
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028826421.1 XP_028682254.1 1739 149
XM_028826422.1 XP_028682255.1 1653 149
XM_028826423.1 XP_028682256.1 1618 149
XM_028826424.1 XP_028682257.1 1716 149
XM_028826425.1 XP_028682258.1 1621 149
XM_028826426.1 XP_028682259.1 1661 149
XM_028826427.1 XP_028682260.1 1619 149
XM_028826428.1 XP_028682261.1 1334 149
XM_028826429.1 XP_028682262.1 1657 149
XM_028826430.1 XP_028682263.1 1600 149

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

surfactant

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline