This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh03644752_g1
See other NSUN5 GE Assays ›
Gene Symbol:
NSUN5
Gene Name:
NOP2/Sun RNA methyltransferase 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 45650479 - 45655970 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh03644752_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NSUN5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001110343.4 XP_001110343.2 825 61
XM_015133312.2 XP_014988798.1 825 61
XM_028845670.1 XP_028701503.1 896 61
XM_028845671.1 XP_028701504.1 896 61
GenBank mRNA JU326324.1 - 710 61
JU476341.1 - 765 61
JV047599.1 - 760 61
JV655722.1 - 695 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0