This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh03986755_s1
See other GPBAR1 GE Assays ›
Gene Symbol:
GPBAR1
Gene Name:
G protein-coupled bile acid receptor 1
Gene Aliases:
EGK_04787
Chromosome Location:
Chr.12: 105661261 - 105666639 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh03986755_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPBAR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260786.1 NP_001247715.1 1043 98
XM_015110963.2 XP_014966449.2 3920 98
XM_015110964.2 XP_014966450.2 4264 98
XM_015110965.2 XP_014966451.2 4040 98
XM_015110967.2 XP_014966453.2 3509 98
XM_015110968.2 XP_014966454.2 3439 98
XM_015110969.2 XP_014966455.2 2887 98
XM_028830417.1 XP_028686250.1 4134 98
XM_028830418.1 XP_028686251.1 3953 98
XM_028830419.1 XP_028686252.1 3917 98
XM_028830420.1 XP_028686253.1 4267 98
XM_028830421.1 XP_028686254.1 1870 98
GenBank mRNA JU470725.1 - 1065 98
JV044656.1 - 1043 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline