Assay Details
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145052.1 | NP_001138524.1 | 454 | 96 | ||
GenBank mRNA | FJ527014.1 | - | 454 | 96 | ||
JU471179.1 | - | 586 | 96 | |||
JV045356.1 | - | 568 | 96 |
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU320406.1 | - | 636 | 96 |
Gene Symbol: LOC106992298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126633.2 | XP_014982119.1 | 754 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
