This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04256356_g1
See other RNF167 GE Assays ›
Gene Symbol:
RNF167
Gene Name:
ring finger protein 167
Gene Aliases:
-
Chromosome Location:
Chr.16: 4787491 - 4792664 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04256356_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RNF167
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194101.2 NP_001181030.2 582 93
XM_015118291.2 XP_014973777.2 744 93
XM_015118292.2 XP_014973778.2 716 93
XM_028835565.1 XP_028691398.1 644 93
XM_028835566.1 XP_028691399.1 498 93
XM_028835567.1 XP_028691400.1 496 93
XM_028835568.1 XP_028691401.1 862 93
GenBank mRNA JU333897.1 - 529 93
JU475081.1 - 529 93
JV046698.1 - 529 93
JV528818.1 - 520 93
JV647793.1 - 535 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0