This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04344418_g1
See other BCS1L GE Assays ›
Gene Symbol:
BCS1L
Gene Name:
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases:
EGK_04799
Chromosome Location:
Chr.12: 106090928 - 106095079 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04344418_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCS1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257781.1 NP_001244710.1 514 65
XM_015110978.2 XP_014966464.1 700 65
XM_015110979.2 XP_014966465.1 658 65
XM_015110980.2 XP_014966466.1 641 65
XM_028830452.1 XP_028686285.1 324 65
XM_028830453.1 XP_028686286.1 255 65
GenBank mRNA JU321170.1 - 514 65
JU470539.1 - 516 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s9s29:80/100.66.79.146:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0