This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh04344721_m1
See other RBFOX3 GE Assays ›
Gene Symbol:
RBFOX3
Gene Name:
RNA binding fox-1 homolog 3
Gene Aliases:
HRNBP3
Chromosome Location:
Chr.16: 75411667 - 75935537 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh04344721_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: RBFOX3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001260861.1
NP_001247790.1
982
109
XM_015120358.2
XP_014975844.1
1207
109
XM_015120360.2
XP_014975846.1
1746
109
XM_015120364.2
XP_014975850.1
1929
109
XM_015120365.2
XP_014975851.1
1510
109
XM_015120367.2
XP_014975853.1
1369
109
XM_028835579.1
XP_028691412.1
1134
109
XM_028835580.1
XP_028691413.1
1137
109
XM_028835581.1
XP_028691414.1
1044
109
XM_028835582.1
XP_028691415.1
996
109
XM_028835583.1
XP_028691416.1
993
109
XM_028835584.1
XP_028691417.1
990
109
XM_028835585.1
XP_028691418.1
996
109
XM_028835586.1
XP_028691419.1
1599
109
XM_028835587.1
XP_028691420.1
903
109
XM_028835588.1
XP_028691421.1
1649
109
XM_028835589.1
XP_028691422.1
1394
109
XM_028835590.1
XP_028691423.1
1513
109
XM_028835591.1
XP_028691424.1
1341
109
XM_028835592.1
XP_028691425.1
1942
109
XM_028835593.1
XP_028691426.1
1355
109
XM_028835594.1
XP_028691427.1
903
109
XM_028835595.1
XP_028691428.1
1649
109
XM_028835596.1
XP_028691429.1
1392
109
XM_028835597.1
XP_028691430.1
1648
109
XM_028835598.1
XP_028691431.1
1510
109
XM_028835599.1
XP_028691432.1
1296
109
XM_028835600.1
XP_028691433.1
1391
109
XM_028835601.1
XP_028691434.1
1507
109
XM_028835602.1
XP_028691435.1
849
109
XM_028835603.1
XP_028691436.1
1508
109
XM_028835604.1
XP_028691437.1
1437
109
XM_028835605.1
XP_028691438.1
1505
109
XM_028835606.1
XP_028691439.1
1422
109
GenBank mRNA
JU320416.1
-
1050
109
JV043894.1
-
982
109
JV565488.1
-
1067
109
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0