This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04344861_m1
See other MOB3B GE Assays ›
Gene Symbol:
MOB3B
Gene Name:
MOB kinase activator 3B
Gene Aliases:
EGK_07640, MOBKL2B
Chromosome Location:
Chr.15: 56226266 - 56427967 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04344861_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261049.1 NP_001247978.1 540 78
XM_015117393.2 XP_014972879.1 885 78
XM_015117395.2 XP_014972881.1 863 78
XM_015117396.2 XP_014972882.1 638 78
GenBank mRNA JU320336.1 - 839 78
JU472134.1 - 540 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Macaca mulatta

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0