This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh04345253_m1
See other GTDC1 GE Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 31101398 - 31479109 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh04345253_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001261488.2
NP_001248417.1
1528
86
XM_015110023.2
XP_014965509.2
3391
86
XM_015110026.2
XP_014965512.2
2980
86
XM_015110027.2
XP_014965513.2
3450
86
XM_015110028.2
XP_014965514.2
2745
86
XM_015110029.2
XP_014965515.2
3170
86
XM_015110031.2
XP_014965517.2
3181
86
XM_015110032.2
XP_014965518.2
2545
86
XM_015110034.2
XP_014965520.2
3256
86
XM_015110035.2
XP_014965521.2
3143
86
XM_015110036.2
XP_014965522.2
3081
86
XM_015110037.2
XP_014965523.2
1627
86
XM_015110038.2
XP_014965524.2
2951
86
XM_015110039.2
XP_014965525.2
2450
86
XM_015110040.2
XP_014965526.2
2305
86
XM_028830336.1
XP_028686169.1
2767
86
XM_028830337.1
XP_028686170.1
4282
86
XM_028830338.1
XP_028686171.1
2266
86
XM_028830340.1
XP_028686173.1
1915
86
XM_028830341.1
XP_028686174.1
1382
86
XM_028830342.1
XP_028686175.1
1491
86
XM_028830343.1
XP_028686176.1
1027
86
XM_028830344.1
XP_028686177.1
1210
86
GenBank mRNA
JU323243.1
-
1574
86
JU471427.1
-
1530
86
JV043889.1
-
1551
86
JV566708.1
-
1463
86
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
3' Most
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-lwb2x:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0