This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04345307_g1
See other ELOVL1 GE Assays ›
Gene Symbol:
ELOVL1
Gene Name:
ELOVL fatty acid elongase 1
Gene Aliases:
EGK_00661
Chromosome Location:
Chr.1: 181099596 - 181104210 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04345307_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELOVL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261559.1 NP_001248488.1 444 90
XM_015136294.2 XP_014991780.1 843 90
XM_015136308.2 XP_014991794.1 712 90
XM_015136315.2 XP_014991801.1 642 90
XM_015136322.2 XP_014991808.1 576 90
XM_028834923.1 XP_028690756.1 586 90
XM_028835005.1 XP_028690838.1 530 90
GenBank mRNA JU335954.1 - 437 90
JU471805.1 - 430 90
JV045630.1 - 444 90
JV566269.1 - 333 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0