This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04346176_m1
See other NDUFC2 GE Assays ›
Gene Symbol:
NDUFC2
Gene Name:
NADH:ubiquinone oxidoreductase subunit C2
Gene Aliases:
EGK_06715
Chromosome Location:
Chr.14: 71148808 - 71159381 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04346176_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266174.1 NP_001253103.1 419 129
GenBank mRNA JU334644.1 - 378 129
JU334645.1 - 378 129
JU475705.1 - 493 129
JU475706.1 - 493 129
JV047151.1 - 419 129
JV545930.1 - 480 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0