This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04347541_s1
See other CMTR2 GE Assays ›
Gene Symbol:
CMTR2
Gene Name:
cap methyltransferase 2
Gene Aliases:
FTSJD1
Chromosome Location:
Chr.20: 60220476 - 60227666 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04347541_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CMTR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261079.1 NP_001248008.1 2814 100
XM_015126533.2 XP_014982019.2 3223 100
XM_015126534.2 XP_014982020.2 3289 100
XM_015126535.2 XP_014982021.2 3239 100
XM_028840548.1 XP_028696381.1 3265 100
XM_028840549.1 XP_028696382.1 2841 100
GenBank mRNA JU335431.1 - 2823 100
JU335432.1 - 2810 100
JU473187.1 - 2814 100
JV704044.1 - 2827 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0