This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04373640_g1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
-
Chromosome Location:
Chr.19: 5604031 - 5613192 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04373640_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015122576.2 XP_014978062.2 1168 107
XM_028839743.1 XP_028695576.1 1186 107
XM_028839744.1 XP_028695577.1 1147 107
XM_028839745.1 XP_028695578.1 1132 107
XM_028839746.1 XP_028695579.1 1129 107
XM_028839749.1 XP_028695582.1 903 107
XM_028839750.1 XP_028695583.1 825 107
XM_028839751.1 XP_028695584.1 864 107
XM_028839752.1 XP_028695585.1 864 107
XM_028839753.1 XP_028695586.1 825 107
XM_028839754.1 XP_028695587.1 974 107
XM_028839755.1 XP_028695588.1 893 107
XM_028839756.1 XP_028695589.1 1192 107
XM_028839757.1 XP_028695590.1 1049 107
XM_028839758.1 XP_028695591.1 1129 107
XM_028839759.1 XP_028695592.1 1100 107
XM_028839761.1 XP_028695594.1 1080 107
XM_028839762.1 XP_028695595.1 1017 107
GenBank mRNA JV581034.1 - 922 107
JV683190.1 - 858 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0