This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04382842_m1
See other CDH18 GE Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
EGK_16383
Chromosome Location:
Chr.6: 19586429 - 20701656 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04382842_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001088306.4 XP_001088306.1 1995 67
XM_015139757.2 XP_014995243.1 2228 67
XM_015139758.2 XP_014995244.1 2215 67
XM_015139759.2 XP_014995245.1 1909 67
XM_015139760.2 XP_014995246.1 2111 67
XM_015139762.2 XP_014995248.1 1869 67
XM_015139763.2 XP_014995249.1 1913 67
XM_028849351.1 XP_028705184.1 1840 67
XM_028849352.1 XP_028705185.1 1875 67
XM_028849353.1 XP_028705186.1 1981 67
XM_028849354.1 XP_028705187.1 2117 67
XM_028849355.1 XP_028705188.1 1909 67
XM_028849356.1 XP_028705189.1 1909 67
GenBank mRNA JU321328.1 - 1869 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

cadherin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0