Assay Details
| Gene Symbol: FOXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834053.1 | XP_028689886.1 | 731 | 61 | ||
| XM_028834054.1 | XP_028689887.1 | 480 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXR1| Gene Symbol: FOXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834053.1 | XP_028689886.1 | 731 | 61 | ||
| XM_028834054.1 | XP_028689887.1 | 480 | 61 | |||
Set Membership: |
|