This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04390082_m1
See other GPER1 GE Assays ›
Gene Symbol:
GPER1
Gene Name:
G protein-coupled estrogen receptor 1
Gene Aliases:
EGK_13399, GPER, GPR30
Chromosome Location:
Chr.3: 34152141 - 34159826 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04390082_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001084531.4 XP_001084531.1 975 60
GenBank mRNA JV465679.1 - 85 60
JV681262.1 - 176 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001084531.4 transcripts.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0