This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04946746_m1
See other GSTT1 GE Assays ›
Gene Symbol:
GSTT1
Gene Name:
glutathione S-transferase theta 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 28154874 - 28164799 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04946746_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318181.1 NP_001305110.1 555 71
XM_015149871.2 XP_015005357.2 534 71
XM_015149872.2 XP_015005358.1 460 71
XM_015149873.2 XP_015005359.2 1378 71
XM_028827229.1 XP_028683062.1 1567 71
XM_028827230.1 XP_028683063.1 656 71
XM_028827231.1 XP_028683064.1 566 71
XM_028827232.1 XP_028683065.1 550 71
XM_028827233.1 XP_028683066.1 372 71
GenBank mRNA JU322564.1 - 584 71
KC734510.1 - 555 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0